Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027892.2 | NP_082168.1 | 22, 23 | 2904 | ||
GenBank mRNA | AK004785.1 | - | 22, 23 | 2904 | ||
AK030460.1 | - | 12, 13 | 1335 | |||
AK035230.1 | - | 14, 15 | 1691 | |||
AK162199.1 | - | 11, 12 | 1336 | |||
BC054076.1 | - | 8, 9 | 2189 | |||
BC125381.1 | - | 23, 24 | 2973 | 40129896 | ||
BC137630.1 | - | 23, 24 | 2973 | 8860649 |