Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019632.3 | NP_062606.1 | 9 | 764 | ||
GenBank mRNA | AK083184.1 | - | 9 | 765 | ||
BC038362.1 | - | 9 | 738 | 5368175 | ||
BC049874.1 | - | 9 | 811 | 5360432 | ||
X61455.1 | - | 8 | 555 |