Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081445.1 | NP_001074914.1 | 14 | 2242 | ||
NM_001113204.1 | NP_001106675.1 | 14 | 2242 | |||
NM_010875.3 | NP_035005.2 | 14 | 2242 | |||
GenBank mRNA | AK051197.1 | - | 14 | 2099 | ||
AK082621.1 | - | 14 | 2095 | |||
BC062942.1 | - | 14 | 2044 | |||
D13289.1 | - | 1 | 24 | |||
X06328.1 | - | 3 | 319 | |||
X15049.1 | - | 14 | 2063 | |||
Y00051.1 | - | 14 | 2063 |