Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133666.3 | NP_598427.1 | 7 | 1144 | ||
GenBank mRNA | AK005021.1 | - | 7 | 1092 | ||
AK075692.1 | - | 7 | 1089 | |||
AK151455.1 | - | 7 | 1055 | |||
AK187850.1 | - | 3 | 332 | |||
AK188080.1 | - | 2 | 227 | |||
AK189786.1 | - | 2 | 333 | |||
AK205680.1 | - | 2 | 205 | |||
AK209435.1 | - | 3 | 332 | |||
BC014818.1 | - | 7 | 1114 | 4456931 | ||
BC041682.1 | - | 7 | 1047 | 4221001 |