Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010890.3 | NP_035020.2 | 3 | 460 | ||
GenBank mRNA | AK050517.1 | - | 3 | 326 | ||
AK054450.1 | - | 2 | 1466 | |||
AK080349.1 | - | 3 | 326 | |||
AK088620.1 | - | 3 | 364 | |||
AK088767.1 | - | 3 | 373 | |||
AK122203.1 | - | 3 | 303 | |||
AK133838.1 | - | 3 | 373 | |||
AK137705.1 | - | 2 | 1476 | |||
AK144240.1 | - | 3 | 374 | |||
AK160570.1 | - | 3 | 325 | |||
AK169582.1 | - | 3 | 370 | |||
BC080710.1 | - | 3 | 243 | |||
BC138813.1 | - | 3 | 338 | 8861835 | ||
BY059586.1 | - | 3 | 348 | |||
D85414.1 | - | 3 | 460 | |||
U96635.1 | - | 3 | 531 |