Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010890.3 | NP_035020.2 | 23 | 2336 | ||
GenBank mRNA | AK088620.1 | - | 23 | 2240 | ||
AK088767.1 | - | 23 | 2249 | |||
AK122203.1 | - | 23 | 2179 | |||
AK133838.1 | - | 23 | 2249 | |||
AK144240.1 | - | 23 | 2249 | |||
BC007184.1 | - | 3 | 183 | |||
BC127070.1 | - | 59 | ||||
BC138813.1 | - | 23 | 2214 | 8861835 | ||
D85414.1 | - | 23 | 2336 | |||
U96635.1 | - | 23 | 2407 |