Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293637.1 | NP_001280566.1 | 3 | 777 | ||
NM_001293638.1 | NP_001280567.1 | 3 | 777 | |||
NM_001293639.1 | NP_001280568.1 | 3 | 777 | |||
NM_175089.4 | NP_780298.2 | 3 | 777 | |||
GenBank mRNA | AK077047.1 | - | 4 | 448 | ||
AK173292.1 | - | 3 | 775 | |||
AY850065.1 | - | 3 | 750 | |||
BC070443.1 | - | 3 | 720 | |||
BC157939.1 | - | 3 | 705 | 9088119 | ||
BC158009.1 | - | 3 | 705 | 9007317 | ||
BC172091.1 | - | 3 | 705 | 9054377 |