Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252250.1 | NP_001239179.1 | 10 | 1493 | ||
NM_001252251.1 | NP_001239180.1 | 10 | 1493 | |||
NM_001252252.1 | NP_001239181.1 | 10 | 1493 | |||
NM_001252253.1 | NP_001239182.1 | 10 | 958 | |||
NM_010898.4 | NP_035028.2 | 10 | 1493 | |||
GenBank mRNA | AK017343.1 | - | 10 | 959 | ||
AK045998.1 | - | 10 | 1315 | |||
AK167685.1 | - | 7 | 584 | |||
BB839081.1 | - | 1 | 22 | |||
BC005442.1 | - | 10 | 1359 | 3496053 | ||
DV654856.1 | - | 7 | 551 | |||
L22987.1 | - | 10 | 951 | |||
L22988.1 | - | 10 | 951 | |||
L22989.1 | - | 10 | 951 | |||
L27090.1 | - | 10 | 944 | |||
L27105.1 | - | 10 | 944 | |||
L28176.1 | - | 10 | 1251 | |||
X74671.1 | - | 10 | 1510 |