Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252250.1 | NP_001239179.1 | 1, 2 | 663 | ||
NM_001252251.1 | NP_001239180.1 | 1, 2 | 663 | |||
NM_001252252.1 | NP_001239181.1 | 1, 2 | 663 | |||
NM_010898.4 | NP_035028.2 | 1, 2 | 663 | |||
GenBank mRNA | AK045998.1 | - | 1, 2 | 485 | ||
BC005442.1 | - | 1, 2 | 529 | 3496053 | ||
CN715904.1 | - | 1, 2 | 221 | |||
L22987.1 | - | 1, 2 | 121 | |||
L22988.1 | - | 1, 2 | 121 | |||
L22989.1 | - | 1, 2 | 121 | |||
L27090.1 | - | 1, 2 | 114 | |||
L27105.1 | - | 1, 2 | 114 | |||
L28176.1 | - | 1, 2 | 421 | |||
X74671.1 | - | 1, 2 | 680 |