Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081453.1 | NP_001074922.1 | 15 | 2134 | ||
NM_001286079.2 | NP_001273008.1 | 16 | 2316 | |||
NM_001286080.2 | NP_001273009.1 | 17 | 2363 | |||
NM_008697.4 | NP_032723.2 | 17 | 2363 | |||
GenBank mRNA | AK129394.1 | - | 17 | 2253 | ||
AK134860.1 | - | 16 | 2329 | |||
AK141891.1 | - | 16 | 2304 | |||
AK162150.1 | - | 17 | 2307 | |||
AK167264.1 | - | 14 | 3350 | |||
AY515727.1 | - | 15 | 2113 | |||
BC137789.1 | - | 16 | 2201 | 8860808 | ||
BC144913.1 | - | 17 | 2248 | 9053461 | ||
BC144914.1 | - | 17 | 2248 | |||
U40342.1 | - | 17 | 2276 |