Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010918.2 | NP_035048.3 | 13 | 2041 | ||
GenBank mRNA | AK035582.1 | - | 12 | 1867 | ||
AK144214.1 | - | 13 | 1952 | |||
L04289.2 | - | 13 | 2046 |