Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nktr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010918.2 | NP_035048.3 | 4, 5 | 381 | ||
GenBank mRNA | AK035582.1 | - | 3, 4 | 282 | ||
AK088056.1 | - | 4, 5 | 320 | |||
AK144214.1 | - | 4, 5 | 292 | |||
AK163916.1 | - | 4, 5 | 394 | |||
BC048414.1 | - | 3, 4 | 315 | |||
L04289.2 | - | 4, 5 | 386 |