Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221628.1 | - | 5 | 1617 | ||
AF191211.1 | - | 5 | 1617 | |||
BC068150.1 | - | 7 | 2204 | |||
BC128307.1 | - | 5 | 1612 | |||
BC128308.1 | - | 5 | 1613 |