Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010928.2 | NP_035058.2 | 20, 21 | 3497 | ||
GenBank mRNA | AK155137.1 | - | 2, 3 | 299 | ||
AK155666.1 | - | 2, 3 | 299 | |||
D32210.1 | - | 20, 21 | 3491 | |||
X68279.1 | - | 15, 16 | 2385 |