Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008716.2 | NP_032742.1 | 33 | 6779 | ||
GenBank mRNA | X74760.1 | - | 33 | 6779 |