Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notch3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008716.2 | NP_032742.1 | 5 | 755 | ||
GenBank mRNA | X74760.1 | - | 5 | 755 |