Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Notch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010929.2 | NP_035059.2 | 22, 23 | 4268 | ||
GenBank mRNA | AK143484.1 | - | 5, 6 | 1433 | ||
BC138044.1 | - | 22, 23 | 4226 | 8861063 | ||
BC145114.1 | - | 23, 24 | 4352 | |||
M80456.1 | - | 22, 23 | 4233 |