Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166371.1 | NP_001159843.1 | 6 | 2274 | ||
NM_008717.3 | NP_032743.2 | 6 | 2284 | |||
GenBank mRNA | AK157791.1 | - | 6 | 2265 | ||
AK163476.1 | - | 5 | 1880 | |||
AK163955.1 | - | 6 | 2236 | |||
AK165189.1 | - | 6 | 2255 | |||
BC076615.1 | - | 6 | 2237 | 30640388 | ||
D83033.1 | - | 6 | 2270 |