Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008727.5 | NP_032753.5 | 10 | 2073 | ||
GenBank mRNA | AK135008.1 | - | 1 | 16 | ||
BC110659.1 | - | 10 | 1953 | 30847317 | ||
BC139767.1 | - | 10 | 1932 | |||
J05504.1 | - | 11 | 2057 | |||
L31932.1 | - | 10 | 1721 |