Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010945.2 | NP_035075.2 | 23, 24 | 2193 | ||
GenBank mRNA | AF013632.1 | - | 23, 24 | 2186 | ||
AK049887.1 | - | 23, 24 | 2145 | |||
AK147709.1 | - | 23, 24 | 2054 | |||
AK164305.1 | - | 23, 24 | 2195 | |||
BC066075.1 | - | 22, 23 | 3088 | |||
BC082810.1 | - | 22, 23 | 3088 | |||
BC145961.1 | - | 23, 24 | 2007 | 40131264 | ||
BC145963.1 | - | 23, 24 | 2007 | 40131266 |