Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmaf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010945.2 | NP_035075.2 | 25 | 2294 | ||
GenBank mRNA | AF013632.1 | - | 25 | 2287 | ||
AK049887.1 | - | 25 | 2246 | |||
AK147709.1 | - | 25 | 2155 | |||
AK164305.1 | - | 25 | 2296 | |||
BC066075.1 | - | 24 | 3189 | |||
BC082810.1 | - | 24 | 3189 | |||
BC145961.1 | - | 25 | 2108 | 40131264 | ||
BC145963.1 | - | 25 | 2108 | 40131266 |