Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010946.3 | NP_035076.1 | 2, 3 | 263 | ||
GenBank mRNA | AK041112.1 | - | 2, 3 | 220 | ||
BC030172.1 | - | 2, 3 | 261 | 5037501 | ||
BY168976.1 | - | 2, 3 | 225 | |||
U57692.1 | - | 2, 3 | 217 |