Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025074.2 | NP_001020245.1 | 12 | 2039 | ||
NM_001282961.1 | NP_001269890.1 | 12 | 2006 | |||
NM_008745.3 | NP_032771.1 | 12 | 2006 | |||
GenBank mRNA | AB377224.1 | - | 11 | 1416 | ||
AB377225.1 | - | 11 | 1416 | |||
AK009606.1 | - | 11 | 1393 | |||
AK018789.1 | - | 12 | 1937 | |||
AK028827.1 | - | 12 | 1859 | |||
AK147391.1 | - | 12 | 1837 | |||
AK147441.1 | - | 11 | 1827 | |||
AK160789.1 | - | 12 | 2007 | |||
BC052014.1 | - | 11 | 1801 | 5707891 | ||
M33385.1 | - | 11 | 1378 | |||
X17647.1 | - | 1889 |