Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025074.2 | NP_001020245.1 | 8 | 1398 | ||
NM_001282961.1 | NP_001269890.1 | 8 | 1365 | |||
NM_008745.3 | NP_032771.1 | 8 | 1365 | |||
GenBank mRNA | AB377224.1 | - | 7 | 775 | ||
AB377225.1 | - | 7 | 775 | |||
AK018789.1 | - | 8 | 1296 | |||
AK028827.1 | - | 8 | 1218 | |||
AK147391.1 | - | 8 | 1196 | |||
AK147441.1 | - | 7 | 1186 | |||
AK160789.1 | - | 8 | 1366 | |||
BC052014.1 | - | 7 | 1160 | 5707891 | ||
KC855567.1 | - | 2 | 104 | |||
M33385.1 | - | 7 | 737 | |||
X17647.1 | - | 1248 |