Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008747.2 | NP_032773.2 | 1 | 144 | ||
GenBank mRNA | AB041826.1 | - | 1 | 120 | ||
AB041827.1 | - | 1 | 120 | |||
AK158943.1 | - | 1 | 144 | |||
BC141019.1 | - | 1 | 139 | 9055631 | ||
U51908.1 | - | 1 | 140 |
Set Membership: |
G-protein coupled receptor |