Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016978.2 | NP_058674.1 | 6, 7 | 921 | ||
GenBank mRNA | AK128914.1 | - | 6, 7 | 912 | ||
AK145877.1 | - | 6, 7 | 922 | |||
AK146413.1 | - | 6, 7 | 866 | |||
AK168851.1 | - | 6, 7 | 870 | |||
BC008119.1 | - | 6, 7 | 854 | 3498240 | ||
X64837.1 | - | 5, 6 | 895 |