Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011020.3 | NP_035150.3 | 6, 7 | 732 | ||
GenBank mRNA | AK144225.1 | - | 6, 7 | 769 | ||
AK163459.1 | - | 7, 8 | 904 | |||
BC057002.1 | - | 6, 7 | 732 | 5702602 | ||
BC110662.1 | - | 6, 7 | 752 | 4511154 | ||
U23921.1 | - | 6, 7 | 849 |