Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016776.2 | NP_058056.2 | 12, 13 | 1734 | ||
GenBank mRNA | AK083908.1 | - | 12, 13 | 1760 | ||
AK143469.1 | - | 12, 13 | 1758 | |||
AK143487.1 | - | 12, 13 | 1767 | |||
AK149747.1 | - | 7, 8 | 1043 | |||
AK155074.1 | - | 12, 13 | 1745 | |||
AK186425.1 | - | 1, 2 | 66 | |||
BC004078.1 | - | 2, 3 | 218 | |||
BC015070.1 | - | 12, 13 | 1735 | |||
BC048858.1 | - | 12, 13 | 1734 | 5363740 | ||
BC052889.1 | - | 12, 13 | 1755 | 30057169 | ||
BC060167.1 | - | 12, 13 | 1740 | |||
U63648.1 | - | 12, 13 | 1743 |