Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2rx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008771.3 | NP_032797.3 | 11 | 1214 | ||
GenBank mRNA | AK007650.1 | - | 5 | 581 | ||
AK035304.1 | - | 11 | 1215 | |||
BC015084.1 | - | 11 | 1228 | 4189541 | ||
X84896.1 | - | 11 | 1208 |