Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P4ha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011030.2 | NP_035160.1 | 6 | 803 | ||
GenBank mRNA | AK030574.1 | - | 6 | 735 | ||
AK045008.1 | - | 6 | 732 | |||
AK149127.1 | - | 6 | 695 | |||
AK160798.1 | - | 6 | 732 | |||
AK165468.1 | - | 6 | 732 | |||
AK169726.1 | - | 6 | 736 | |||
BC009654.1 | - | 6 | 726 | 3484815 | ||
U16162.1 | - | 5 | 573 |