Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pax4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159925.1 | NP_001153397.1 | 3 | 392 | ||
NM_001159926.1 | NP_001153398.1 | 3 | 392 | |||
NM_011038.2 | NP_035168.1 | 3 | 392 | |||
GenBank mRNA | AB008912.1 | - | 3 | 367 | ||
AB010557.1 | - | 3 | 335 | |||
AB010558.1 | - | 3 | 350 | |||
AF031150.1 | - | 3 | 425 | |||
AF104231.1 | - | 2 | 147 | |||
AF145233.1 | - | 3 | 339 | |||
BC131906.1 | - | 3 | 303 | 40130183 | ||
BC131908.1 | - | 3 | 301 | 40130185 | ||
BC145168.1 | - | 3 | 303 | 9053719 | ||
BC145169.1 | - | 3 | 303 | 9053720 | ||
Y09584.1 | - | 1 | 31 |