Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcsk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008793.2 | NP_032819.1 | 6 | 678 | ||
GenBank mRNA | AK161504.1 | - | 5 | 760 | ||
D01093.1 | - | 6 | 678 |