Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 7 | 1020 | ||
GenBank mRNA | AK051425.1 | - | 7 | 1021 | ||
AK141083.1 | - | 1 | 39 | |||
AK143518.1 | - | 7 | 1048 | |||
AK145483.1 | - | 7 | 1050 | |||
AK166188.1 | - | 7 | 1229 | |||
AK180401.1 | - | 1 | 73 | |||
AK180999.1 | - | 1 | 73 | |||
AK194254.1 | - | 1 | 73 | |||
AK194908.1 | - | 1 | 73 | |||
AK203854.1 | - | 1 | 73 | |||
AK214881.1 | - | 2 | 136 | |||
BC011069.1 | - | 7 | 763 | 4006745 | ||
BC013663.1 | - | 7 | 1079 | 4208974 | ||
X69025.1 | - | 7 | 1042 |