Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011049.4 | NP_035179.1 | 6, 7 | 943 | ||
GenBank mRNA | AK051425.1 | - | 6, 7 | 944 | ||
AK143518.1 | - | 6, 7 | 971 | |||
AK145483.1 | - | 6, 7 | 973 | |||
AK166188.1 | - | 6, 7 | 1152 | |||
AK214881.1 | - | 1, 2 | 59 | |||
BC011069.1 | - | 6, 7 | 686 | 4006745 | ||
BC013663.1 | - | 6, 7 | 1002 | 4208974 | ||
X69025.1 | - | 6, 7 | 965 |