Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pctp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008796.2 | NP_032822.2 | 4 | 467 | ||
GenBank mRNA | AF114437.1 | - | 2 | 172 | ||
AF151639.1 | - | 4 | 438 | |||
BB614866.1 | - | 315 | ||||
BC053478.1 | - | 422 | ||||
BC071234.1 | - | 4 | 423 | |||
Z50024.1 | - | 4 | 296 |