Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011053.2 | NP_035183.2 | 7 | 876 | ||
GenBank mRNA | AK129080.1 | - | 7 | 874 | ||
AK141450.1 | - | 7 | 877 | |||
AK147950.1 | - | 7 | 861 | |||
AK161803.1 | - | 7 | 866 | |||
BC010844.1 | - | 7 | 856 | |||
BC070468.1 | - | 7 | 863 | 30628812 |