Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008803.2 | NP_032829.1 | 16 | 1546 | ||
GenBank mRNA | AF067806.1 | - | 1612 | |||
AK148238.1 | - | 1755 | ||||
BC125578.1 | - | 16 | 1546 | 40130093 | ||
BC132145.1 | - | 16 | 1546 | 40130422 |