Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008806.2 | NP_032832.2 | 4 | 871 | ||
GenBank mRNA | AK039397.1 | - | 4 | 583 | ||
AK044364.1 | - | 4 | 814 | |||
BC048247.1 | - | 4 | 850 | |||
BC058803.1 | - | 4 | 818 | |||
BC129923.1 | - | 4 | 801 | 40111882 | ||
BC129924.1 | - | 4 | 800 | |||
X55968.1 | - | 4 | 804 | |||
X60133.1 | - | 4 | 811 | |||
X87952.1 | - | 4 | 811 |