Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pepd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008820.2 | NP_032846.2 | 7, 8 | 576 | ||
GenBank mRNA | AK010581.1 | - | 7, 8 | 557 | ||
AK145843.1 | - | 7, 8 | 607 | |||
AK168384.1 | - | 7, 8 | 566 | |||
AK206317.1 | - | 2, 3 | 102 | |||
BC086644.1 | - | 7, 8 | 550 | 6807359 | ||
BC131945.1 | - | 7, 8 | 578 | 40130222 | ||
BC131973.1 | - | 7, 8 | 578 | 40130250 | ||
D82983.1 | - | 7, 8 | 563 | |||
U51014.1 | - | 7, 8 | 599 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK183750.1 | - | 278 | Chr. - - [-] on Build |