Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008823.3 | NP_032849.2 | 5 | 780 | ||
GenBank mRNA | AK150212.1 | - | 5 | 741 | ||
AK152782.1 | - | 5 | 731 | |||
AK154306.1 | - | 5 | 765 | |||
AK159291.1 | - | 5 | 781 | |||
AK159609.1 | - | 5 | 731 | |||
AK159853.1 | - | 5 | 731 | |||
AK171374.1 | - | 5 | 764 | |||
BC132369.1 | - | 5 | 764 | 40130646 | ||
BC138306.1 | - | 5 | 764 | 8861326 | ||
X12905.1 | - | 4 | 629 |