Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011074.2 | NP_035204.2 | 4 | 561 | ||
GenBank mRNA | AF033655.1 | - | 4 | 520 | ||
AK016370.1 | - | 4 | 503 | |||
AK045083.1 | - | 4 | 562 | |||
AK051283.1 | - | 4 | 535 | |||
AK051411.1 | - | 4 | 504 | |||
AK083269.1 | - | 3 | 716 | |||
AK083790.1 | - | 4 | 504 | |||
AK087398.1 | - | 3 | 332 | |||
AK129226.1 | - | 4 | 388 | |||
AK138437.1 | - | 3 | 802 | |||
AK139386.1 | - | 3 | 722 | |||
AK141535.1 | - | 3 | 490 | |||
AK165149.1 | - | 3 | 406 | |||
BC068134.1 | - | 3 | 765 | 6834018 | ||
U62391.1 | - | 3 | 415 |