Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045558.1 | NP_001039023.1 | 3, 4 | 395 | ||
NM_008837.3 | NP_032863.2 | 3, 4 | 454 | |||
GenBank mRNA | AK050224.1 | - | 3, 4 | 366 | ||
AK090155.1 | - | 387 | ||||
AK162724.1 | - | 3 | 320 | |||
BC023088.1 | - | 3, 4 | 361 | |||
BC048788.1 | - | 3, 4 | 340 | 6488048 | ||
BU057774.1 | - | 3, 4 | 357 | |||
BU698586.1 | - | 3, 4 | 453 | |||
CA468666.1 | - | 3, 4 | 275 | |||
CK791908.1 | - | 3, 4 | 395 | |||
Z46720.1 | - | 2, 3 | 143 |