Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pick1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045558.1 | NP_001039023.1 | 11 | 1072 | ||
NM_008837.3 | NP_032863.2 | 11 | 1131 | |||
GenBank mRNA | AK050224.1 | - | 8 | 1897 | ||
AK090155.1 | - | 1064 | ||||
BC023088.1 | - | 10 | 1667 | |||
BC030363.1 | - | 3 | 1376 | |||
BC030880.1 | - | 2 | 1207 | |||
BC030912.1 | - | 2 | 283 | |||
BC040824.1 | - | 2 | 1207 | |||
BC048788.1 | - | 11 | 1017 | 6488048 | ||
BE652516.1 | - | 3 | 267 | |||
CD363874.1 | - | 8 | 561 | |||
Z46720.1 | - | 10 | 820 |