Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3c2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011083.2 | NP_035213.2 | 13 | 2579 | ||
GenBank mRNA | AK142284.1 | - | 14 | 2608 | ||
BC096553.1 | - | 12 | 2013 | |||
U52193.1 | - | 14 | 2485 | |||
U55772.1 | - | 12 | 1895 |