Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip5k1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008846.2 | NP_032872.1 | 5 | 436 | ||
GenBank mRNA | AK133809.1 | - | 5 | 431 | ||
AK146100.1 | - | 5 | 436 | |||
AK149354.1 | - | 5 | 436 | |||
AK164552.1 | - | 5 | 419 | |||
BC034864.1 | - | 5 | 450 | 3326897 | ||
D86176.1 | - | 4 | 369 |