Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008850.1 | NP_032876.1 | 9 | 915 | ||
GenBank mRNA | AK144618.1 | - | 10 | 928 | ||
AK149745.1 | - | 10 | 905 | |||
AK151496.1 | - | 10 | 904 | |||
AK152089.1 | - | 10 | 928 | |||
AK152874.1 | - | 10 | 922 | |||
AK153569.1 | - | 10 | 928 | |||
AK165659.1 | - | 10 | 920 | |||
AK167760.1 | - | 9 | 928 | |||
AK168187.1 | - | 9 | 885 | |||
AK168720.1 | - | 10 | 918 | |||
AK183653.1 | - | 3 | 179 | |||
AK186336.1 | - | 106 | ||||
AK187601.1 | - | 3 | 178 | |||
AK207617.1 | - | 3 | 178 | |||
BC056171.1 | - | 10 | 870 | 5710926 | ||
S72681.1 | - | 9 | 690 | |||
U96725.1 | - | 9 | 916 |