Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011103.3 | NP_035233.1 | 2 | 254 | ||
GenBank mRNA | AB011812.1 | - | 1 | 17 | ||
AF251036.1 | - | 1 | 17 | |||
AK144231.1 | - | 2 | 259 | |||
AK168768.1 | - | 2 | 249 | |||
AK171476.1 | - | 2 | 255 | |||
AY545076.1 | - | 1 | 17 | |||
BC051416.1 | - | 2 | 218 | 6515302 | ||
HQ840432.1 | - | 1 | 17 | |||
M69042.1 | - | 2 | 249 | |||
X60304.1 | - | 1 | 30 |