Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011103.3 | NP_035233.1 | 10 | 1153 | ||
GenBank mRNA | AB011812.1 | - | 9 | 916 | ||
AB201453.1 | - | 7 | 755 | |||
AB201454.1 | - | 7 | 755 | |||
AB201455.1 | - | 7 | 755 | |||
AB201456.1 | - | 7 | 755 | |||
AF251036.1 | - | 9 | 916 | |||
AK144231.1 | - | 10 | 1158 | |||
AK168768.1 | - | 10 | 1148 | |||
AK171476.1 | - | 10 | 1155 | |||
AY545076.1 | - | 9 | 916 | |||
BC051416.1 | - | 10 | 1117 | 6515302 | ||
M69042.1 | - | 10 | 1148 | |||
X60304.1 | - | 9 | 929 |