Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008861.3 | NP_032887.3 | 14 | 2732 | ||
GenBank mRNA | AF014010.1 | - | 14 | 2635 | ||
AK053502.1 | - | 14 | 2637 | |||
AK128961.1 | - | 15 | 2506 | |||
BC053058.1 | - | 14 | 2739 | |||
BC062969.1 | - | 14 | 2739 | |||
CX566027.1 | - | 5 | 459 | |||
FJ609779.1 | - | 13 | 2480 | |||
Y13278.1 | - | 14 | 2648 |