Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008861.3 | NP_032887.3 | 14 | 2802 | ||
GenBank mRNA | AF014010.1 | - | 14 | 2705 | ||
AK053502.1 | - | 14 | 2707 | |||
AK128961.1 | - | 15 | 2576 | |||
AK217384.1 | - | 1 | 15 | |||
BC053058.1 | - | 14 | 2809 | |||
BC062969.1 | - | 14 | 2809 | |||
CX566027.1 | - | 5 | 529 | |||
FJ609779.1 | - | 13 | 2550 | |||
Y13278.1 | - | 14 | 2718 |